Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent BioTek High Flow Complete Dispense/Waste System
GSA_VA

Catalog No. BT2120533 Shop All Agilent Technologies Inc. Products
Click to view available options
Capacity (Metric):
4 L
10 L
20 L

High Flow Complete Dispense/Waste System for use with the 406 FX Washer Dispenser.

  • Includes four dispense bottles, two waste bottles and high flow vacuum pump

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 10 L
Includes 4 x 10 L Dispense Bottles, 2 x 10 L Waste Bottles, High Flow Vacuum Pump
Type Waste System
For Use With (Equipment) 406 FX Washer Dispenser
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.