Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ COMP/Thrombospondin-5 Antibody Pair [Biotin]
SDP

Catalog No. p-7183044 Shop All Novus Biologicals Products
Click to view available options
Quantity:
15 Plates
5 Plates

Polyclonal/Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes an antibody specific for Human COMP/Thrombospondin-5.

Specifications

Assay Range 39.1 pg/mL
Assay Sensitivity 39.06-2500 pg/mL
Assay Sandwich ELISA
Conjugate Biotin
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Human COMP/Thrombospondin-5.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias cartilage oligomeric matrix protein, cartilage oligomeric matrix protein (pseudoachondroplasia, epiphyseal dysplasia1, multiple), cartilage oligomeric matrix protein(pseudoachondroplasia, epiphyseal dysplasia1, multiple), EDM1, EPD1, MEDMGC131819, MGC149768, PSACH, pseudoachondroplasia (epiphyseal dysplasia 1, multiple), THBS5, Thrombospondin-5, TSP5
Gene ID (Entrez) 1311
Gene Symbol COMP
Host Species Rabbit/Mouse
Kit Contents
  • Standard: Each vial contains 60 ng of recombinant COMP/Thrombospondin-5.
  • Capture Antibody: 0.5 mg/mL of mouse anti-COMP/Thrombospondin-5 monoclonal antibody.
  • Detection Antibody: Each vial contains 120 μg biotinylated rabbit anti-COMP/Thrombospondin-5 polyclonal antibody. Reconstitute with sterile 1 mL distilled water.
  • Quantity 15 Plates
    Regulatory Status RUO
    Storage Requirements Storage is content dependent.
    Target COMP/Thrombospondin-5
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to a working concentration of 2 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 2500 pg/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.