Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement C3a Antibody (D17/1), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP223497A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP223497A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP223497A32 Supplier Novus Biologicals Supplier No. NBP223497AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Complement C3a Monoclonal specifically detects Complement C3a in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA.

Specifications

Antigen Complement C3a
Applications Western Blot, ELISA
Classification Monoclonal
Clone D17/1
Conjugate Alexa Fluor 532
Dilution Western Blot, ELISA, Sandwich ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product
Gene Symbols C3
Host Species Mouse
Immunogen Human C3a
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 718
Test Specificity Recognizes an epitope that is present on human C3, C3a and C3a (desArg). Does not cross-react with C4a or C5a.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.