Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement C3a Antibody (K13/16-5.7) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB018311 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018311 0.025 mg
NBP105122 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018311 Supplier Novus Biologicals Supplier No. NBP1051220.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Complement C3a Monoclonal specifically detects Complement C3a in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Sandwich ELISA Capture.

Specifications

Antigen Complement C3a
Applications Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, ELISA
Classification Monoclonal
Clone K13/16-5.7
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:10000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:75, Immunohistochemistry-Paraffin 1:150, Sandwich ELISA Capture
Formulation 10mM Phosphate (pH 7.4) and 0.5M NaCl with 0.05% Sodium Azide
Gene Alias AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product
Gene Symbols C3
Host Species Mouse
Immunogen Human C3a
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 718
Test Specificity Specific for human C3, C3a and C3a (desArg) and does not cross-react with C4a or C5a.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.