Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement C3b Antibody (2B10B9B2), Novus Biologicals™
SDP

Catalog No. NBP149635 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP149635 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP149635 Supplier Novus Biologicals Supplier No. NBP149635
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Complement C3b Monoclonal specifically detects Complement C3b in Rat samples. It is validated for Western Blot, Immunoassay.

Specifications

Antigen Complement C3b
Applications Western Blot, Immunoassay
Classification Monoclonal
Clone 2B10B9B2
Concentration 0.1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunoassay
Gene Alias AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product
Gene Symbols C3
Host Species Mouse
Immunogen The monoclonal antibody 2B10B9B2 reacts with an epitope on C3b.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 718
Target Species Rat
Content And Storage Store at 4°C.
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.