Learn More
Description
Specifications
Specifications
| Antigen | Complement C3 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 4C6A8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product |
| Gene Symbols | C3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
