Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement C3d Mouse anti-Human, Clone: C3D/2891, Novus Biologicals™
SDP

Catalog No. NB122956 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB122956 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB122956 Supplier Novus Biologicals Supplier No. NBP30774120UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Complement C3d Monoclonal specifically detects Complement C3d in Human samples. It is validated for ELISA, Protein Array.

Specifications

Antigen Complement C3d
Applications ELISA, Peptide Array
Classification Monoclonal
Clone C3D/2891
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution ELISA, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Alias AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product
Host Species Mouse
Immunogen Recombinant full-length human Complement C3d protein
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 718
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.