Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement C4 Antibody, Novus Biologicals™
SDP

Catalog No. NB186445 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB186445 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB186445 Supplier Novus Biologicals Supplier No. NBP312295
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Complement C4 Polyclonal antibody specifically detects Complement C4 in Human, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen Complement C4
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:10000, Immunohistochemistry 1:100, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin 1:100
Formulation Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide
Gene Alias C3 and PZP-like alpha-2-macroglobulin domain-containing protein 2, C4, complement component 4A (Rodgers blood group), RG
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 1220-1270 on human complement C4-A preprotein.
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 720
Target Species Human, Primate
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.