Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement C4 Antibody, Novus Biologicals™
SDP

Catalog No. NB119899 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119899 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119899 Supplier Novus Biologicals Supplier No. NBP311274
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Complement C4 Polyclonal specifically detects Complement C4 in Rat samples. It is validated for Immunoprecipitation, Immunoassay.

Specifications

Antigen Complement C4
Applications Immunoprecipitation, Immunoassay
Classification Polyclonal
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Immunoprecipitation, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias C3 and PZP-like alpha-2-macroglobulin domain-containing protein 2, C4, complement component 4A (Rodgers blood group), RG
Host Species Rabbit
Immunogen The polyclonal antibody reacts with Rat C4.
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 720
Target Species Rat
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.