Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement C4b Antibody (25.3F4) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP231554 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP231554 100 μg
NB019503 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP231554 Supplier Novus Biologicals Supplier No. NBP231554

Mouse Monoclonal Antibody

Complement C4b Monoclonal specifically detects Complement C4b in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Complement C4b
Applications Western Blot, ELISA
Classification Monoclonal
Clone 25.3F4
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Gene Alias basic C4, Basic complement C4, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 3, C4B1, C4B12, C4B2, C4B3, C4FMGC164979, CH, Chido form of C4, CO4C4B5, complement C4-B, complement C4B1a, complement component 4B, complement component 4B (Chido blood group), CPAMD3FLJ60561, EC 2.1.1.144, EC 2.7.11
Gene Symbols C4B
Host Species Mouse
Immunogen C4 isolated from human plasma adsorbed onto aluminum hydroxide gel.
Molecular Weight of Antigen 204 kDa
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 721
Test Specificity Epitope specificity differs from that of NBP1-05084, as determined by inhibition ELISA.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.