Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement C4c Antibody (99-72-18), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB154817 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB154817 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB154817 Supplier Novus Biologicals Supplier No. NBP312091DL594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Complement C4c Monoclonal antibody specifically detects Complement C4c in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence

Specifications

Antigen Complement C4c
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 99-72-18
Conjugate DyLight 594
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias basic C4, Basic complement C4, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 3, C4B1, C4B12, C4B2, C4B3, C4FMGC164979, CH, Chido form of C4, CO4C4B5, complement C4-B, complement C4B1a, complement component 4B, complement component 4B (Chido blood group), CPAMD3FLJ60561, EC 2.1.1.144, EC 2.7.11
Host Species Mouse
Immunogen Synthetic peptide consisting of the last 15C-terminal residues of the C4c a4-domain (Cys-EELVYELNPLDHRGR).
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 721
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.