Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement C4d Antibody (12D11), Novus Biologicals™
SDP

Catalog No. NBP262802 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP262802 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP262802 Supplier Novus Biologicals Supplier No. NBP262802
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Complement C4d Monoclonal antibody specifically detects Complement C4d in Human, Porcine, Bovine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoassay.

Specifications

Antigen ComplementC4d
Applications Flow Cytometry, Immunohistochemistry (Frozen), In Situ Hybridization (ISH), Western Blot
Classification Polyclonal
Clone 12D11
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay
Formulation PBS with No Preservative
Gene Symbols C4A
Host Species Mouse
Immunogen Compliment factor C4D
Purification Method Protein G purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) 720
Target Species Human, Pig, Bovine, Primate, Monkey
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.