Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement C5 Antibody (568), Novus Biologicals™
SDP

Catalog No. NB12011879 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12011879 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB12011879 Supplier Novus Biologicals Supplier No. NB12011879
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Complement C5 Monoclonal specifically detects Complement C5 in Human samples. It is validated for Western Blot, Immunoassay.

Specifications

Antigen Complement C5
Applications Western Blot, Immunoassay
Classification Monoclonal
Clone 568
Concentration 0.1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunoassay
Gene Alias anaphylatoxin C5a analog, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4, C5, complement C5, complement component 5, CPAMD4MGC142298, FLJ17816, FLJ17822
Gene Symbols C5
Host Species Mouse
Immunogen Unfortunately, this information is considered to be commercially sensitive.
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 727
Test Specificity This reacts with an epitope on C5 and on C5b.
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.