Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Component C5a Antibody (C17/5), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP223498A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP223498A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP223498A94 Supplier Novus Biologicals Supplier No. NBP223498AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Complement Component C5a Monoclonal antibody specifically detects Complement Component C5a in Human, Baboon samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Complement Component C5a
Applications Western Blot, ELISA
Classification Monoclonal
Clone C17/5
Conjugate Alexa Fluor 594
Dilution Western Blot, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias C5, C5A, HE
Gene Symbols C5
Host Species Mouse
Immunogen Purified human C5a
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 727
Test Specificity Clone C17/5 reacts with human C5a/C5a(desArg) and cross-reacts with baboon C5a/C5a(desArg).
Target Species Human, Baboon
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.