Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Component C5a Antibody (C17/5) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB019304 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mg
0.025 mL
0.1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019304 0.025 mL
NBP223498 0.25 mg
NBP22349801 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB019304 Supplier Novus Biologicals Supplier No. NBP2234980.025ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Complement C5 Monoclonal specifically detects Complement C5 in Human, Baboon samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Complement C5
Applications Western Blot, ELISA
Classification Monoclonal
Clone C17/5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Formulation 10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide
Gene Alias anaphylatoxin C5a analog, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4, C5, complement C5, complement component 5, CPAMD4MGC142298, FLJ17816, FLJ17822
Gene Symbols C5
Host Species Mouse
Immunogen Purified human C5a
Purification Method Protein A or G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 727
Test Specificity Clone C17/5 reacts with human C5a/C5a(desArg) and cross-reacts with baboon C5a/C5a(desArg).
Target Species Baboon, Human
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.