Learn More
Description
Specifications
Specifications
| Antigen | Complement C5 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:10 |
| Formulation | 0.2 um filtered solution in PBS and 0.1% BSA with 0.02% Sodium Azide |
| Gene Accession No. | P01031 |
| Gene Alias | anaphylatoxin C5a analog, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4, C5, complement C5, complement component 5, CPAMD4MGC142298, FLJ17816, FLJ17822 |
| Gene Symbols | C5 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
