Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Complement C5a Monoclonal Antibody (D4), Invitrogen™

Catalog No. p-200020169
Change view
Click to view available options
Quantity:
20 μL
200 μL
Catalog No. Quantity
FEMA550990 20 μL
FEMA550989 200 μL
2 options

Catalog No. FEMA550990

Supplier: Invitrogen™ MA550990

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The protein encoded by this gene is the fifth component of complement, which plays an important role in inflammatory and cell killing processes. This protein is comprised of alpha and beta polypeptide chains that are linked by a disulfide bridge. An activation peptide, C5a, which is an anaphylatoxin that possesses potent spasmogenic and chemotactic activity, is derived from the alpha polypeptide via cleavage with a convertase. The C5b macromolecular cleavage product can form a complex with the C6 complement component, and this complex is the basis for formation of the membrane attack complex, which includes additional complement components. Mutations in this gene cause complement component 5 deficiency, a disease where patients show a propensity for severe recurrent infections. Defects in this gene have also been linked to a susceptibility to liver fibrosis and to rheumatoid arthritis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement C5a
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone D4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene C5
Gene Accession No. P01031, P08650
Gene Alias anaphylatoxin; anaphylatoxin C5a analog; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4; C4B1; C4B12; C4B2; C4B3; C4B5; C4F; C5; C5a; C5a anaphylatoxin; C5b; C5D; CH; CO4; complement C5; Complement C5 alpha chain; Complement C5 alpha' chain; Complement C5 beta chain; complement component 5; CPAMD3; CPAMD4; DADB-112B14.11; ECLZB; FLJ17816; FLJ17822; FLJ60561; Hc; He; Hemolytic complement; MGC142298; MGC164979; prepro-C5; RGD1561905
Gene Symbols C5
Host Species Mouse
Immunogen Recombinant Human C5a protein, Thr678-Arg751 (Accession #P01031).
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362119, 727
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.