Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement C6 Antibody (3G11), Novus Biologicals™
SDP

Catalog No. NB119815 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119815 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119815 Supplier Novus Biologicals Supplier No. NBP311190
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Complement C6 Monoclonal specifically detects Complement C6 in Rat samples. It is validated for Western Blot, Immunohistochemistry-Frozen, Immunoassay.

Specifications

Antigen Complement C6
Applications Western Blot, Immunohistochemistry (Frozen), Immunoassay
Classification Monoclonal
Clone 3G11
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry-Frozen, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias complement component 6, complement component C6
Host Species Mouse
Immunogen The monoclonal antibody 3G11 was raised by immunization of mice with C6 isolated from rat serum.
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 729
Target Species Rat
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.