Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement C6 Antibody, Novus Biologicals™
SDP

Catalog No. NB12011899 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12011899 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB12011899 Supplier Novus Biologicals Supplier No. NB12011899
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Complement C6 Polyclonal specifically detects Complement C6 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen Complement C6
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation 0.2 um filtered solution in PBS and 0.1% BSA with 0.02% Sodium Azide
Gene Alias complement component 6, complement component C6
Gene Symbols C6
Host Species Rabbit
Immunogen Unfortunately, this information is considered to be commercially sensitive
Purification Method IgG purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 729
Test Specificity recognises C6.
Target Species Mouse
Content And Storage Store at 4°C.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.