Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement C7 Antibody (113), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB334199 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB334199 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB334199 Supplier Novus Biologicals Supplier No. NBP290244MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Complement C7 Monoclonal antibody specifically detects Complement C7 in Human samples. It is validated for ELISA

Specifications

Antigen Complement C7
Applications ELISA
Classification Monoclonal
Clone 113
Conjugate mFluor Violet 610 SE
Dilution Sandwich ELISA Capture
Formulation 50mM Sodium Borate
Gene Alias complement component 7, complement component C7
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Human Complement C7 (Uniprot#: P10643, Met1-Gln843)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 730
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.