Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement C7 Rabbit anti-Human, Clone: 113, Novus Biologicals™
SDP

Catalog No. NB021952 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB021952 50 μg
NB021951 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB021952 Supplier Novus Biologicals Supplier No. NBP29024450UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Complement C7 Monoclonal specifically detects Complement C7 in Human samples. It is validated for Sandwich ELISA Capture.

Specifications

Antigen Complement C7
Applications ELISA
Classification Monoclonal
Clone 113
Conjugate Unconjugated
Dilution Sandwich ELISA Capture 1:250-1:2000
Formulation 0.2 um filtered solution in PBS
Gene Alias complement component 7, complement component C7
Host Species Rabbit
Immunogen Recombinant Human Complement C7 Protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 730
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.