Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement C8A/B/G Antibody (056B-373) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10064340 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064340 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064340 Supplier Novus Biologicals Supplier No. NB10064340
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 3 publications

Complement C8A/B/G Monoclonal specifically detects Complement C8A/B/G in Human samples. It is validated for ELISA, Flow Cytometry, IF/IHC, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot.

Specifications

Antigen Complement C8A/B/G
Applications ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 056B-373
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Gene Accession No. P07360
Gene Alias complement component 8, alpha polypeptide, complement component C8 alpha chain
Gene Symbols C8A
Host Species Mouse
Immunogen Purified human C8
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 731
Test Specificity This antibody recognizes complement component 8 (C8), a 151 kDa member of the complement C6/C7/C8/C9 family, present in blood serum. C8 is a terminal component of the complement system, part of both the complement membrane attack complex (MAC), and important to MAC assembly. C8 binds to the C5b-7 complex, anchored to the membrane, creating C5b-8. C5b-8 binds C9 and catalyzes the polymerization of C9 molecules to form C5-b9 (MAC). C8 is thought to contain lipid binding sites, facilitating the insertion of MAC into the membrane. Defects in the alpha chain of C8 can result in complement C8 deficiency type I. Furthermore, C8 deficiencies can cause recurring bacterial infections, in particular from Neisseria meningitides.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.