Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Component C1r Antibody (OTI1F1), HRP, Novus Biologicals™
SDP

Catalog No. NBP270318H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270318H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270318H Supplier Novus Biologicals Supplier No. NBP270318H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Complement Component C1r Monoclonal specifically detects Complement Component C1r in Human, Canine samples. It is validated for Western Blot.

Specifications

Antigen Complement Component C1r
Applications Western Blot
Classification Monoclonal
Clone OTI1F1
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias complement C1r subcomponent, Complement component 1 subcomponent r, complement component 1, r subcomponent, EC 3.4.21, EC 3.4.21.41
Gene Symbols C1R
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 262-562 of human Complement Component C1r (NP_001724) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 715.0
Target Species Human, Canine
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.