Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Component C1s Antibody (9), FITC, Novus Biologicals™
SDP

Catalog No. NBP105023F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105023F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP105023F Supplier Novus Biologicals Supplier No. NBP105023F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Complement Component C1s Monoclonal specifically detects Complement Component C1s in Human samples. It is validated for ELISA, Immunohistochemistry.

Specifications

Antigen Complement Component C1s
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 9
Conjugate FITC
Dilution ELISA, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias basic proline-rich peptide IB-1, C1 esterase, complement C1s subcomponent, Complement component 1 subcomponent s, complement component 1, s subcomponent, EC 3.4.21, EC 3.4.21.42, FLJ44757
Gene Symbols C1S
Host Species Mouse
Immunogen C1s proenzyme purified from human plasma
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 716
Test Specificity Specific for C1s proenzyme and activated free C1s
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.