Learn More
Complement Component C3aR Antibody (3G7), Alexa Fluor™ 750 - Chimeric, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Complement Component C3aR |
| Applications | Flow Cytometry, ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 3G7 |
| Conjugate | Alexa Fluor 750 |
| Dilution | Flow Cytometry, ELISA, Immunofluorescence |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | AZ3B, AZ3BC3a anaphylatoxin chemotactic receptor, C3a-R, C3AR, C3AR1, C3ARcomplement component 3 receptor 1, C3R1, complement component 3a receptor 1, Complement Component C3a R, Complement Component C3aR, HNFAG09 |
| Gene Symbols | C3AR1 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.