Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Component C3aR Antibody (620613), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB340030 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340030 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340030 Supplier Novus Biologicals Supplier No. FAB10417MFV450
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Complement Component C3aR Monoclonal specifically detects Complement Component C3aR in Mouse samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Complement Component C3aR
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 620613
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias AZ3B, AZ3BC3a anaphylatoxin chemotactic receptor, C3a-R, C3AR, C3AR1, C3ARcomplement component 3 receptor 1, C3R1, complement component 3a receptor 1, Complement Component C3a R, Complement Component C3aR, HNFAG09
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived transfected with mouse C3aR, Accession # O09047
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 719
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.