Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Component C3aR Antibody (hC3aRZ1), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB10064835J Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064835J 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064835J Supplier Novus Biologicals Supplier No. NB10064835APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Complement Component C3aR Monoclonal specifically detects Complement Component C3aR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen).

Specifications

Antigen Complement Component C3aR
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone hC3aRZ1
Conjugate APC
Dilution Flow Cytometry
Gene Accession No. Q16581
Gene Alias AZ3B, AZ3BC3a anaphylatoxin chemotactic receptor, C3a-R, C3AR, C3AR1, C3ARcomplement component 3 receptor 1, C3R1, complement component 3a receptor 1, Complement Component C3a R, Complement Component C3aR, HNFAG09
Gene Symbols C3AR1
Host Species Mouse
Immunogen Recombinant human C3aR protein
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 719
Test Specificity NB100-64835 recognizes the receptor for human complement component C3a (C3aR), a 55kD cell surface protein of the rhodopsin family of G protein-coupled seven-transmembrane receptors. C3a is an anaphylatoxin, which is a potent proinflammatory mediator that possesses immunoregulatory properties. C3a regulates inflammatory functions by binding to the C3a receptor. C3aR is expressed on various tissues and leucocytes including basophils, neutrophils, mast cells, eosinophils and activated T lymphocytes.
Expression levels of C3aR on human cells varies significantly between donors.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.