Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Component C5/C5a Antibody (06), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB353202 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB353202 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB353202 Supplier Novus Biologicals Supplier No. NBP306182MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Complement Component C5/C5a Monoclonal antibody specifically detects Complement Component C5/C5a in Human samples. It is validated for ELISA

Specifications

Antigen Complement Component C5/C5a
Applications ELISA
Classification Monoclonal
Clone 06
Conjugate mFluor Violet 610 SE
Dilution Sandwich ELISA Capture
Formulation 50mM Sodium Borate
Gene Alias C5a;C5b;C5D;Complement C5;CPAMD4;ECLZB
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Complement Component C5/C5a (Accession#: NP_001726.2; Leu679-Arg751).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 727
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.