Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Component C5a Antibody (215), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB108403 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB108403 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB108403 Supplier Novus Biologicals Supplier No. NBP289578AF594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Complement Component C5a Monoclonal specifically detects Complement Component C5a in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Complement Component C5a
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 215
Conjugate Alexa Fluor 594
Formulation 50 mM Sodium Borate
Gene Alias C5, C5A, HE
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Complement Component C5a (CO5_HUMAN; Leu679-Arg751).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 727
Test Specificity Has cross-reactivity in ELISA with: Human A2M
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.