Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Component C5a Antibody (C17/5), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB355668 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355668 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355668 Supplier Novus Biologicals Supplier No. NBP223498MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Complement Component C5a Monoclonal antibody specifically detects Complement Component C5a in Human, Baboon samples. It is validated for Western Blot, ELISA

Specifications

Antigen Complement Component C5a
Applications Western Blot, ELISA
Classification Monoclonal
Clone C17/5
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias C5, C5A, HE
Host Species Mouse
Immunogen Purified human C5a
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 727
Target Species Human, Baboon
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.