Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Component C5aR1 Antibody (347234), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB371914 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB371914 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB371914 Supplier Novus Biologicals Supplier No. FAB36481D
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Complement Component C5aR1 Monoclonal specifically detects Complement Component C5aR1 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry, Immunohistochemistry.

Specifications

Antigen Complement Component C5aR1
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone 347234
Conjugate DyLight 350
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias C5A, C5a anaphylatoxin chemotactic receptor, C5aR, C5a-R, C5ARC5a anaphylatoxin receptor, C5R1C5a ligand, CD88, CD88 antigen, complement component 5 receptor 1, complement component 5 receptor 1 (C5a ligand), complement component 5a receptor 1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Complement Component C5aR1, Met1-Val350, Accession # P21730
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 728
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.