Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Component C5aR1 Antibody (583837) [Janelia Fluor™ 646], Novus Biologicals™
SDP

Catalog No. NB376551 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB376551 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB376551 Supplier Novus Biologicals Supplier No. FAB6467J
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Complement Component C5aR1 Monoclonal antibody specifically detects Complement Component C5aR1 in Mouse samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen Complement Component C5aR1
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 583837
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias C5A, C5a anaphylatoxin chemotactic receptor, C5aR, C5a-R, C5ARC5a anaphylatoxin receptor, C5R1C5a ligand, CD88, CD88 antigen, complement component 5 receptor 1, complement component 5 receptor 1 (C5a ligand), complement component 5a receptor 1
Host Species Rat
Immunogen NS0 mouse myeloma cell line transfected with mouse Complement Component C5a R1, Accession # P30993
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 728
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.