Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Component C5aR2 Antibody (1D9-M12), Novus Biologicals™
SDP

Catalog No. NB180416 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB180416 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB180416 Supplier Novus Biologicals Supplier No. NBP307018
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Complement Component C5aR2 Monoclonal antibody specifically detects Complement Component C5aR2 in Human, Primate samples. It is validated for Western Blot, Flow Cytometry

Specifications

Antigen Complement Component C5aR2
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 1D9-M12
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry 2 to 6 μg/mL
Formulation PBS
Gene Alias C5a anaphylatoxin chemotactic receptor C5L2, C5L2G-protein coupled receptor 77, G protein-coupled receptor C5L2, GPF77, GPR77
Host Species Mouse
Immunogen L1.2 cells transfected with human C5aR2
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 27202
Target Species Human, Primate
Content And Storage Store at 4°CC.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.