Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Component C9 Antibody (22), HRP, Novus Biologicals™
SDP

Catalog No. NBP221612H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP221612H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP221612H Supplier Novus Biologicals Supplier No. NBP221612H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Complement Component C9 Monoclonal specifically detects Complement Component C9 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen Complement Component C9
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 22
Conjugate HRP
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias complement component 9, complement component C9
Gene Symbols C9
Host Species Mouse
Immunogen Complement factor C9 adsorbed onto aluminum hydroxide gel.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 735
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.