Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Component C9 Antibody (C9-6-25-5), Novus Biologicals™
SDP

Catalog No. NB119834 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119834 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119834 Supplier Novus Biologicals Supplier No. NBP311209
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Complement Component C9 Monoclonal specifically detects Complement Component C9 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Complement Component C9
Applications Western Blot
Classification Monoclonal
Clone C9-6-25-5
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias complement component 9, complement component C9
Host Species Rat
Immunogen Murine C9 peptide
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 735
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.