Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Factor B Antibody (M13/12), Novus Biologicals™
SDP

Catalog No. NBP149638 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP149638 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP149638 Supplier Novus Biologicals Supplier No. NBP149638
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Complement Factor B Monoclonal specifically detects Complement Factor B in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunoassay.

Specifications

Antigen Complement Factor B
Applications Western Blot, Immunoprecipitation, Immunoassay
Classification Monoclonal
Clone M13/12
Concentration 0.1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500, Immunoassay
Gene Alias BF AHUS4, B-factor, properdin, BFD, C3 proaccelerator, C3 proactivator, C3/C5 convertase, complement factor B, EC 3.4.21, EC 3.4.21.47, FB, FBI12, GBGCFAB, Glycine-rich beta glycoprotein, glycine-rich beta-glycoprotein, H2-Bf, PBF2, Properdin factor B
Gene Symbols CFB
Host Species Mouse
Immunogen The monoclonal antibody M13/12 recognizes human complement factor B/Bb.
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 629
Target Species Human
Content And Storage Store at 4°C.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.