Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Factor D/Adipsin Antibody (I8/1) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB018310 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018310 0.025 mg
NBP105121 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018310 Supplier Novus Biologicals Supplier No. NBP1051210.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Complement Factor D/Adipsin Monoclonal specifically detects Complement Factor D/Adipsin in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Complement Factor D/Adipsin
Applications Western Blot, ELISA
Classification Monoclonal
Clone I8/1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:1000
Formulation 10mM Phosphate (pH 7.4) and 0.5M NaCl with 0.05% Sodium Azide
Gene Alias Adipsin, ADNcomplement factor D, C3 convertase activator, complement factor D (adipsin), complement factor D preproprotein, D component of complement (adipsin), DF, EC 3.4.21, EC 3.4.21.46, PFD, Properdin factor DADIPSIN
Gene Symbols CFD
Host Species Mouse
Immunogen Human factor D.
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1675
Test Specificity Clone I8/1 is specific for complement factor D in human serum and plasma
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.