Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Factor D/Adipsin Antibody (I8/1), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP105121G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105121G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP105121G Supplier Novus Biologicals Supplier No. NBP105121G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Complement Factor D/Adipsin Monoclonal specifically detects Complement Factor D/Adipsin in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Complement Factor D/Adipsin
Applications Western Blot, ELISA
Classification Monoclonal
Clone I8/1
Conjugate DyLight 488
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Adipsin, ADNcomplement factor D, C3 convertase activator, complement factor D (adipsin), complement factor D preproprotein, D component of complement (adipsin), DF, EC 3.4.21, EC 3.4.21.46, PFD, Properdin factor DADIPSIN
Gene Symbols CFD
Host Species Mouse
Immunogen Human factor D.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1675
Test Specificity Clone I8/1 is specific for complement factor D in human serum and plasma
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.