Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Factor D/Adipsin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | Complement Factor D/Adipsin |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP179793
![]() |
Novus Biologicals
NBP179793 |
100 μL |
Each for $487.50
|
|
|||||
NBP17979320
![]() |
Novus Biologicals
NBP17979320UL |
20 μL | N/A | N/A | N/A | ||||
Description
Complement Factor D/Adipsin Polyclonal specifically detects Complement Factor D/Adipsin in Human samples. It is validated for Western Blot.Specifications
Complement Factor D/Adipsin | |
Polyclonal | |
Rabbit | |
NP_001919 | |
1675 | |
Synthetic peptide directed towards the C terminal of human CFD. Peptide sequence GRRPDSLQHVLLPVLDRATCNRRTHHDGAITERLMCAESNRRDSCKGDSG. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
Adipsin, ADNcomplement factor D, C3 convertase activator, complement factor D (adipsin), complement factor D preproprotein, D component of complement (adipsin), DF, EC 3.4.21, EC 3.4.21.46, PFD, Properdin factor DADIPSIN | |
CFD | |
IgG | |
24 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title