Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Complement Factor D Monoclonal Antibody (I8/1b)
GREENER_CHOICE

Catalog No. ABG080102
Change view
Click to view available options
Quantity:
400 μL
Catalog No. Quantity
ABG-080102 400 μL
1 options

Catalog No. ABG-080102

Supplier: Invitrogen™ GAU0080102

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GAU 008-01-02 detects Complement factor D in human serum and plasma. GAU 008-01-02 has been successfully used in ELISA and Western blot procedures. Use in Western blotting under non-reducing conditions. GAU 008-01 may be used as detection antibody in sandwich ELISA with GAU 010-04-02 as a capture antibody.

Complement factor D is a serine protease of the alternative pathway of complement activation. The protein is a single chain polypeptide with a molecular mass of 25 kDa. Factor D cleaves factor B bound to C3b, generating the alternative pathway C3 convertase C3bBb and releasing the Ba fragment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement Factor D
Applications ELISA, Western Blot
Classification Monoclonal
Clone I8/1b
Concentration 1.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene CFD
Gene Accession No. P00746
Gene Alias 28 kDa adipocyte protein; adipsin; Adn; C3 convertase activator; Cfd; Complement factor D; complement factor D (adipsin); complement factor D preproprotein; D component (adipsin) of complement; D component of complement (adipsin); Df; EC 3.4.21.46; Endogenous vascular elastase; EVE; PFD; Properdin factor D
Gene Symbols CFD
Host Species Mouse
Immunogen Human complement factor D.
Purification Method Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1675
Target Species Human
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.