Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Factor H Antibody (10-10), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB199645 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB199645 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB199645 Supplier Novus Biologicals Supplier No. NBP252743AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Complement Factor H Monoclonal antibody specifically detects Complement Factor H in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)

Specifications

Antigen Complement Factor H
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 2023-10-10 00:00:00
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias adrenomedullin binding protein, age-related maculopathy susceptibility 1, AHUS1, AMBP1, ARMD4, beta-1H, beta-1-H-globulin, CFHL3, complement factor H, factor H, factor H-like 1, FH, FHL1, H factor 1, H factor 1 (complement), H factor 2 (complement), HF, HF1ARMS1, HF2, HUSMGC88246
Host Species Mouse
Immunogen Purified human factor H from serum.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3075
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.