Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Factor I Antibody (OTI7C9), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP270997A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP270997A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP270997A32 Supplier Novus Biologicals Supplier No. NBP270997AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Complement Factor I Monoclonal specifically detects Complement Factor I in Human samples. It is validated for Western Blot.

Specifications

Antigen Complement Factor I
Applications Western Blot
Classification Monoclonal
Clone OTI7C9
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias AHUS3, C3B/C4B inactivator, C3BINA, C3b-inactivator, complement component I, complement control protein factor I, complement factor I, complement factor I heavy chain, EC 3.4.21, EC 3.4.21.45, FIC3b-INA, I factor (complement), IF, KAF, Konglutinogen-activating factor, light chain of factor I
Gene Symbols CFI
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 340-583 of human Complement Factor I (NP_000195) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3426
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.