Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Factor MASP3 Antibody (MM0197-4F19) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP212204 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212204 0.1 mg
NB019098 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP212204 Supplier Novus Biologicals Supplier No. NBP212204
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Complement Factor MASP3 Monoclonal specifically detects Complement Factor MASP3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Complement Factor MASP3/MASP1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0197-4F19
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-1000, Immunohistochemistry-Paraffin 1:10-1:500
Formulation Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative
Gene Accession No. NP_624302
Gene Alias Complement factor MASP-3, Complement-activating component of Ra-reactive factor, CRARF1, CRARFMAP1, DKFZp686I01199, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.104, EC 3.4.21.42, FLJ26383, mannan-binding lectin serine peptidase 1 (C4/C2 activating component ofRa-reactive factor), mannan-binding lectin serine protease 1, mannan-binding lectin serine protease 1 (C4/C2 activating component ofRa-reactive factor), Mannose-binding lectin-associated serine protease 1, Mannose-binding protein-associated serine protease, MAp44, MASP, MASP-1, MASP3, MGC126283, MGC126284, PRSS5, Ra-reactive factor serine protease p100, RaRF, Serine protease 5
Gene Symbols MASP1
Host Species Mouse
Immunogen Human MASP1
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5648
Reconstitution Reconstitute with sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.