Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Factor MASP3 Antibody (MM0197-4F19), PerCP, Novus Biologicals™
SDP

Catalog No. NB361257 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361257 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361257 Supplier Novus Biologicals Supplier No. NBP212204PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Complement Factor MASP3 Monoclonal antibody specifically detects Complement Factor MASP3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Complement Factor MASP3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0197-4F19
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias Complement-activating component of Ra-reactive factor, CRARF, Mannan-binding lectin serine protease 1 isoform 2, Mannose-binding protein-associated serine protease 3, MASP3, RaRF
Host Species Mouse
Immunogen Human recombinant Complement Factor MASP3 (Catalytic Domain)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5648
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.