Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement receptor C5aR (CD88) Mouse anti-Human, Clone: S5/1, MilliporeSigma™
Mouse Monoclonal Antibody
$192.35 - $452.71
Specifications
Antigen | Complement receptor C5aR (CD88) |
---|---|
Clone | S5/1 |
Concentration | Please refer to lot specific datasheet. |
Applications | ELISA, Flow Cytometry, Immunoprecipitation, Inhibition Assays, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF1980-25
|
MilliporeSigma
MABF198025UL |
25 μL |
Each for $192.35
|
|
|||||
MABF1980-100
|
MilliporeSigma
MABF1980100UL |
100 μL |
Each for $452.71
|
|
|||||
Description
Anti-Complementreceptor C5aR (CD88), clone S5/1 Flow Cytometry, Immunoprecipitation, Inhibition/Activity assays, andn Western Blotting.Specifications
Complement receptor C5aR (CD88) | |
Please refer to lot specific datasheet. | |
Monoclonal | |
Purified | |
RUO | |
Human | |
C5AR1,C5AR,C5R1 | |
IgG2a κ | |
Protein G Purified | |
Clone S5/1 specifically detects C5a anaphylatoxin chemotactic receptor 1 (C5aR). It targets an epitope within the first 31 amino acids from the N-terminal region. |
S5/1 | |
ELISA, Flow Cytometry, Immunoprecipitation, Inhibition Assays, Western Blot | |
Unconjugated | |
Mouse | |
Inflammation and Immunology | |
NP_001727.1 | |
BSA-conjugated linear peptide corresponding to the first 31 amino acids from the N-terminal region of human C5a anaphylatoxin chemotactic receptor 1. | |
Primary | |
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title