Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complexin-2 Rabbit anti-Human, Alexa Fluor 750, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB188130 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB188130 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB188130 Supplier Novus Biologicals Supplier No. NBP314762AF750
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Complexin-2 Polyclonal antibody specifically detects Complexin-2 in Human samples. It is validated for ELISA, Immunofluorescence

Specifications

Antigen Complexin-2
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias complexin 2, Complexin II, complexin-2, CPX II, CPX2, CPX-2, DKFZp547D155, Hfb1, MGC138492, synaphin 1,921-L, synaphin-1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human CPLX2 (Q6PUV4-1; Asp2-Lys134)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10814
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.