Learn More
Description
Highly suited for isolation of cell surface glycoproteins from detergent-solubilized membranes.
- Con A coupled to Sepharose, in a 4% agarose matrix via cyanogen bromide activation
- Binds molecules that contain α-D-mannose, α-D-glucose, and sterically related residues with available C-3, C-4, or C-5 hydroxyl groups
- Group-specific adsorbent for molecules containing sugars
- Concentration of immobilized Con A, approx. 13mg/mL medium; binding capacity for porcine thyroglobulin, approx. 30mg/mL medium
- Stable in the pH range 4 to 9, and in the presence of all commonly used buffers
- Average particle size: 90μm
- Delivered in 0.1M acetate buffer, pH 6.0, containing 1M NaCl, 1mM CaCl2, 1mM MgCl2, 1mM MnCl2, and 20% ethanol
Specifications
Specifications
| Binding Capacity | 30mg/mL |
| Ligand Concentration | 13mg Con A/mL Drained Medium |
| Matrix | 4% Agarose |
| Particle Size | 90 μm |
| pH | 4 to 9 |
| Quantity | 100 mL |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | For separation and purification of Glycoproteins, Polysaccharides and Glycolipids, isolates cell surface Glycoproteins from detergent-solubilized membranes |
| Ligand | Concanavalin A |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.