Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Concanavalin A (Con A) Conjugates

Catalog No. p-7244555
Encompass_Preferred
Click to view available options
Conjugate:
Alexa Fluor 350
Alexa Fluor 488
Alexa Fluor 594
Alexa Fluor 633
Alexa Fluor 647
Alexa Fluor Plus 405
Alexa Fluor Plus 750
Tetramethylrhodamine
Texas Red
Color:
Blue
Deep Red
Green
Near-infrared
Orange
Red
Violet
Excitation/Emission:
346/442 nm
405/450 nm
495/519 nm
555/580 nm
590/617 nm
595/615 nm
632/647 nm
650/668 nm
750/777 nm
Protein Subtype:
Concanavalin A (Con A)
Concanavalin A, succinylated

Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates.

Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates. Concanavalin A (Con A) has been used in a variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC) and flow cytometry (FC). In many cell types, concanavalin A (Con A) is a good marker of ER and Golgi.

Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Flour and Alexa Flour Plus conjugates. Concanavalin A selectively binds to alpha‐mannopyranosyl and alpha‐glucopyranosyl residues. In neutral and alkaline solutions, concanavalin A exists as a tetramer with a molecular weight of approximately 104,000 Daltons. In acidic solutions (pH below 5.0), concanavalin A exists as a dimer. Conjugated concanavalin A has been used in variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC), and flow cytometry (FC).

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Immunocytochemistry, Immunohistochemistry, Immunofluorescence
Quantity 5 mg
Source Canavlia ensiformis
Conjugate Alexa Fluor 350
Cross Reactivity α-man, α-glc
Species C. ensiformis
Recombinant Native
Protein Tag None
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Excitation/Emission 346/442 nm
Label Type Alexa Fluor Dyes
Shipping Condition Room Temperature
Expression System Natural
Protein Family Lectins
Form Lyophilized
Protein Subtype Concanavalin A (Con A)
Research Category Imaging
Color Blue
Product Line Alexa Fluor™
Purity or Quality Grade Imaging Quality
Ligand Type Lectin
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.