Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ Connector Cables for Automated Thermal Cycler (ATC)

Catalog No. p-7246656
Encompass_Preferred
Change view
Click to view available options
Format:
1-m connector cable (flexible)
3-m connector cable (flexible)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
A31482 1-m connector cable (flexible)
A31483 3-m connector cable (flexible)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A31482 Supplier Applied Biosystems™ Supplier No. A31482
Only null left
Add to Cart
Add to Cart

Includes

1 meter connection cable for Automated Thermal Cycler (ATC)

Used to connect the Automated Thermal Cycler (ATC) block to the power box, this connector is available in a rigid 10-cm, flexible 1-m, or flexible 3-m cable format.

For customers seeking to automate their PCR workflow, the Applied Biosystems Automated Thermal Cycler (ATC) provides the reliability and performance you've come to expect from Applied Biosystems instruments. Small, easy-to-integrate, with a compatible format, our Automated Thermal Cycler enables hands-free PCR results.

In order to better align with lab design and demands, a longer or shorter cable may be utilized to optimize workflow.

Warranty and Services

27 month warranty

TRUSTED_SUSTAINABILITY

Specifications

Format 1-m connector cable (flexible)
For Use With (Equipment) Automated Thermal Cycler (ATC)
Product Type Connector Cable
For Use With (Application) PCR
Max. Heating Rate (Thermal Block) 2.8°C/sec. (Block), 1.6°C/sec. (Sample)
Quantity 1 cable
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.