Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connexin 26/GJB2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP241304 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP241304 0.1 mg
NB396410 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP241304 Supplier Novus Biologicals Supplier No. NBP241304
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

Connexin 26/GJB2 Polyclonal specifically detects Connexin 26/GJB2 in Human, Rat, Porcine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Connexin 26/GJB2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 2 ug/mL, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS with 0.02% Sodium Azide
Gene Alias beta 2, 26kD (connexin 26), Cx26, CX26DFNA3, DFNA3A, DFNB1, gap junction protein, beta 2, 26kDa, gap junction protein, beta 2, 26kDa (connexin 26), NSRD1
Gene Symbols GJB2
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid peptide near the center of human GJB2. The immunogen is located within amino acids 80 - 130 of GJB2. Amino Acid Squence: HVAYRRHEKKRKFIKGE
Molecular Weight of Antigen 26 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience, Sensory Systems
Primary or Secondary Primary
Gene ID (Entrez) 2706
Test Specificity GJB2 antibody is human specific.
Target Species Human, Rat, Pig
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.